Gossypin
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
APOSBIDF1030-100MGEA
111
GBP
APOSBIDF1030-100MG
APOSBIDF1030-250MG
APOSBIDF1030-50MG
Gossypin
Gossypin
Storage Temperature:
Ambient |
CAS Number:
652-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...