AC1NUZLU

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Z-Val-Ala-DL-Asp-fluoromethylketone

APOSBIBA1001-5MGEA 379 GBP
APOSBIBA1001-5MG
AC1NUZLU
AC1NUZLU
Formula: C₂₁H₂₈FN₃O₇
MW: 453.467 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06795862
CAS Number: 220644-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR