Nɑ-Acetyl-L-glutamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Ac-Gln-OHN-Acetyl-L-glutamineAc-L-Gln-OHN-Ac-Gln-OH

APOSBIA1740-50GEA 0 GBP
APOSBIA1740-50G APOSBIA1740-100G
Nɑ-Acetyl-L-glutamine
Nalpha-Acetyl-L-glutamine
Formula: C₇H₁₂N₂O₄
MW: 188.18 g/mol
Melting Pt: 206…208 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00038159
CAS Number: 2490-97-3
EINECS: 219-647-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR