Atrazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

BIA0156-5G
APOSBIA0156-5GEA 153 GBP
APOSBIA0156-5G
Atrazine
Atrazine
Formula: C₈H₁₄ClN₅
MW: 215.69 g/mol
Melting Pt: 175 °C
Density: 7.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041810
CAS Number: 1912-24-9
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR