2,3-Difluorostyrene 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Difluoro-3-ethenylbenzene, 1,2-Difluoro-3-vinylbenzene

PC6273-1G PC6273-250MG PC6273-5G
APOSPC6273-1GEA 70.4 GBP
APOSPC6273-1G APOSPC6273-250MG APOSPC6273-5G
2,3-Difluorostyrene 97%
2,3-Difluorostyrene
Formula: C₈H₆F₂
MW: 140.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12827910
CAS Number: 1080636-91-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR