1H,1H-Undecafluoro(2-methyl-3-oxahexan-1-ol) 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H,1H-Perfluoro(2-methyl-3-oxahexan-1-ol), 2-(Heptafluoropropxy)-2,3,3,3-tetrafluoropropan-1-ol

PC6210D-25G PC6210D-5G
APOSPC6210D-25GEA 327 GBP
APOSPC6210D-25G APOSPC6210D-5G
1H,1H-Undecafluoro(2-methyl-3-oxahexan-1-ol) 97%
1H,1H-Undecafluoro(2-methyl-3-oxahexan-1-ol)
Formula: C₆H₃F₁₁O₂
MW: 316.069 g/mol
Boiling Pt: 119…120 °C
Density: 1.553 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236069
CAS Number: 26537-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR