3-Bromo-2-(2,2,2-trifluoroethoxy)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC57065-1G PC57065-5G
APOSPC57065-1GEA 57.9 GBP
APOSPC57065-1G APOSPC57065-5G
3-Bromo-2-(2,2,2-trifluoroethoxy)pyridine
3-Bromo-2-(2,2,2-trifluoroethoxy)pyridine
Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858511
CAS Number: 760207-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR