12,12,12-Trifluorododecane-1-thiol 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC56220-1G PC56220-250MG
APOSPC56220-1GEA 586 GBP
APOSPC56220-1G APOSPC56220-250MG
12,12,12-Trifluorododecane-1-thiol 97%
12,12,12-Trifluorododecane-1-thiol
Formula: C₁₂H₂₃F₃S
MW: 256.371 g/mol
Boiling Pt: 101…104 °C (2 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD28334496
CAS Number: 168703-96-6
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR