3-Fluoro-2-nitrophenylthiocyanate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC508053-1G PC508053-250MG PC508053-5G
APOSPC508053-1GEA 311 GBP
APOSPC508053-1G APOSPC508053-250MG APOSPC508053-5G
3-Fluoro-2-nitrophenylthiocyanate
3-Fluoro-2-nitrophenylthiocyanate
Formula: C₇H₃FN₂O₂S
MW: 198.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723307
CAS Number: 2149598-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR