Hexafluoroisopropyl methyl ether 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Methyl hexafluoroisopropyl ether

PC4757-25G PC4757-5G PC4757-100G
APOSPC4757-25GEA 119 GBP
APOSPC4757-25G APOSPC4757-5G APOSPC4757-100G
Hexafluoroisopropyl methyl ether 99%
Hexafluoroisopropyl methyl ether
Formula: C₄H₄F₆O
MW: 182.064 g/mol
Boiling Pt: 50 °C
Density: 1.39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00221772
CAS Number: 13171-18-1
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR