1,1,1,2,2,5,5,6,6,6-Decafluoro-3-iodohexane
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC450084-1G
PC450084-250MG
APOSPC450084-1GEA
206
GBP
APOSPC450084-1G
APOSPC450084-250MG
1,1,1,2,2,5,5,6,6,6-Decafluoro-3-iodohexane
1,1,1,2,2,5,5,6,6,6-Decafluoro-3-iodohexane
Formula:
C₆H₃F₁₀I MW: 391.977 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28136374 CAS Number: 1980048-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...