3-Amino-N-(4-fluorophenyl)propanoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC1868-1G PC1868-5G
APOSPC1868-1GEA 110 GBP
APOSPC1868-1G APOSPC1868-5G
3-Amino-N-(4-fluorophenyl)propanoic acid
3-Amino-N-(4-fluorophenyl)propanoic acid
Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784304
CAS Number: 38470-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR