4-[(4-Methylhomopiperazin-1-yl)methyl]benzylamine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]benzylamine, 4-[(4-Methyl-1,4-diazepan-1-yl)methyl]benzylamine, {4-[(4-Methyl-1,4-diazepan-1-yl)methyl]phenyl}methylamine

OR9672-1G OR9672-250MG
APOSOR9672-1GEA 357 GBP
APOSOR9672-1G APOSOR9672-250MG
4-[(4-Methylhomopiperazin-1-yl)methyl]benzylamine 95%
4-[(4-Methylhomopiperazin-1-yl)methyl]benzylamine
Formula: C₁₄H₂₃N₃
MW: 233.353 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08435915
CAS Number: 884507-52-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR