4-Chloropyrimidine-2-carbonitrile 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR943779-1G OR943779-250MG
APOSOR943779-1GEA 516 GBP
APOSOR943779-1G APOSOR943779-250MG
4-Chloropyrimidine-2-carbonitrile 95%
4-Chloropyrimidine-2-carbonitrile
Formula: C₅H₂ClN₃
MW: 139.542 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09607718
CAS Number: 898044-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR