(3,3-Dimethylcyclobutyl)methanol
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR922737-1G
OR922737-5G
APOSOR922737-1GEA
426
GBP
APOSOR922737-1G
APOSOR922737-5G
(3,3-Dimethylcyclobutyl)methanol
(3,3-Dimethylcyclobutyl)methanol
Formula:
C₇H₁₄O MW: 114.185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17170398 CAS Number: 75017-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...