2-Isobutanoyl-3-picoline 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR909600-10G OR909600-1G OR909600-5G
APOSOR909600-10GEA 546 GBP
APOSOR909600-10G APOSOR909600-1G APOSOR909600-5G
2-Isobutanoyl-3-picoline 97%
2-Isobutanoyl-3-picoline
Formula: C₁₀H₁₃NO
MW: 163.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16087791
CAS Number: 1247742-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR