3-Nitro-4-(((tetrahydro-2H-pyran-4-yl)methyl)amino)benzenesulphonamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Nitro-3-(oxan-4-ylmethylamino)benzenesulphonamide

OR55052-10G OR55052-1G OR55052-5G
APOSOR55052-10GEA 94.9 GBP
APOSOR55052-10G APOSOR55052-1G APOSOR55052-5G
3-Nitro-4-(((tetrahydro-2H-pyran-4-yl)methyl)amino)benzenesulphonamide
3-Nitro-4-(((tetrahydro-2H-pyran-4-yl)methyl)amino)benzenesulphonamide
Formula: C₁₂H₁₇N₃O₅S
MW: 315.345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28742285
CAS Number: 1228779-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR