2-Bromo-3-nitrobenzamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR50483-1G OR50483-250MG OR50483-5G
APOSOR50483-1GEA 188 GBP
APOSOR50483-1G APOSOR50483-250MG APOSOR50483-5G
2-Bromo-3-nitrobenzamide
2-Bromo-3-nitrobenzamide
Formula: C₇H₅BrN₂O₃
MW: 245.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00086360
CAS Number: 35757-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR