2,5-Dichloro-4-(trimethylstannyl)pyridine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR303173-1G OR303173-500MG
APOSOR303173-1GEA 131 GBP
APOSOR303173-1G APOSOR303173-500MG
2,5-Dichloro-4-(trimethylstannyl)pyridine
2,5-Dichloro-4-(trimethylstannyl)pyridine
Formula: C₈H₁₁Cl₂NSn
MW: 310.796 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689170
CAS Number: 1201787-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR