N-(4-Aminophenyl)-N-phenylbenzamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR303152-1G OR303152-500MG OR303152-5G
APOSOR303152-1GEA 195 GBP
APOSOR303152-1G APOSOR303152-500MG APOSOR303152-5G
N-(4-Aminophenyl)-N-phenylbenzamide
N-(4-Aminophenyl)-N-phenylbenzamide
Formula: C₁₉H₁₆N₂O
MW: 288.343 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689164
CAS Number: 73333-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR