2-(4-Methylphenyl)-1,3,4-oxadiazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(P-tolyl)-1,3,4-oxadiazole

APOSOR111447-1GEA 0 GBP
APOSOR111447-1G
2-(4-Methylphenyl)-1,3,4-oxadiazole
2-(4-Methylphenyl)-1,3,4-oxadiazole
Formula: C₉H₈N₂O
MW: 160.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00489537
CAS Number: 827-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR