2,3-Dihydrobenzo[b]furan-7-carboxaldehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,3-Dihydro-1-benzofuran-7-carboxaldehyde, 2,3-Dihydro-7-formyl-1-benzofuran

OR101068-1G OR101068-250MG
APOSOR101068-1GEA 77.7 GBP
APOSOR101068-1G APOSOR101068-250MG
2,3-Dihydrobenzo[b]furan-7-carboxaldehyde 98%
2,3-Dihydrobenzo[b]furan-7-carboxaldehyde
Formula: C₉H₈O₂
MW: 148.159 g/mol
Melting Pt: 54…55 °C
Storage Temperature: Ambient
MDL Number: MFCD06200853
CAS Number: 196799-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR