Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Sulpho succinimidyl 4-(ρ-maleimidophenyl) butyrate
, Sulfo SMPB
, Sulphosuccinimidyl 4-(N-maleimidophenyl)butyrate
APOSBICL201-100MGEA
153
GBP
APOSBICL201-100MG
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Primary amine and Sulphhydryl reactiveSpacer arm -Water soluble
Ref: Bangs, J.D.et.al. (1986) J.Cell.Biol. 103, 255-263
Formula:
C₁₈H₁₅N₂O₉S·Na MW: 458.374 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00054981 CAS Number: 92921-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...