Ethanol denatured 96%, GPR RECTAPUR® (UK HMRC Spec)

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ethyl alcohol denatured

88377.360EA 89.2 GBP
88377.360
Ethanol denatured 96%, GPR RECTAPUR® (UK HMRC Spec)
Ethanol denatured
Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Appearance - free from visible impurity Passes test
Density (20 °C) 0.802 - 0.812 g/cm³
Alcoholic content (20 °C) 95.7 - 96.1 % v/v

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR