Hydrochloric acid 4 mol/l (4 N) (20°C), Titripur®, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Muriatic acidHydrochloric acid solution

M-Clarity™ quality level = MQ200

1.60328.1003EA 34.3 GBP
1.60328.1003 1.60328.1000
Hydrochloric acid 4 mol/l (4 N) (20°C), Titripur®, Supelco®
Hydrochloric acid

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 3.980 - 4.020 mol/l
Measurement uncertainty +/- 0.012 mol/l
Traceability NIST SRM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR