Tris(4-nitrophenyl)amine, Technical Grade, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

335495-5GEA 66 GBP
335495-5G
Tris(4-nitrophenyl)amine, Technical Grade, Sigma-Aldrich®
Tris(4-nitrophenyl)amine
Formula: C₁₈H₁₂N₄O₆
Melting Pt: >300 °C (lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00024636
CAS Number: 20440-93-1

Order Now

Specification Test Results

Appearance Yellow-green or gold powder
Infrared spectrum Conforms to structure
Proton NMR spectrum Conforms to structure
High pressure liquid Typically ca. 97%
Elemental analysis Carbon typically 54.8% - 58.8%, Nitrogen typically 14.2% - 15.2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR