ANTI-NTRK3 Mouse Monoclonal Antibody [clone: BS337]
Supplier: Biosensis
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-1837-100
BSENM-1837-100EA 294 GBP
BSENM-1837-100
ANTI-NTRK3 Mouse Monoclonal Antibody [clone: BS337]
Antibodies
Receptor tyrosine kinase involved in nervous system and probably heart development. Upon binding of its ligand NTF3/neurotrophin-3, NTRK3 autophosphorylates and activates different signaling pathways, including the phosphatidylinositol 3-kinase/AKT and the MAPK pathways, that control cell survival and differentiation (Ref: uniprot.org).
Applications: Flow Cytometry (5-10 ug/mL): Tested on human and rodent cell lines. Cell staining can be performed under native conditions on ice, or on fixed cells with up to 4% formaldehyde.
Immunocytochemistry (1-2 ug/mL): Tested on fixed (4% formaldehyde) human cells.
Other applications have not been tested. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.