Chemical resistant gloves, double dipped PVC, A845
Supplier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRTWA845GNRXLEA 7.5 GBP
PRTWA845GNRXL
Chemical resistant gloves, double dipped PVC, A845
Gloves
Double dipped PVC gauntlet with sandy palm finish for enhanced grip. Jersey lined for extra wearer comfort.
  • Chemical resistant gauntlet
Certifications: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .10.1, EN ISO 374-5 Micro Organisms (Pass), EN 388:2016 - (3.1.2.1.X), EN ISO 374-1:2016 Type B (J.K.L), EN 420 Dexterity 5.