Desisopropyl atrazine
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Atrazine desisopropyl
EHERC10332000EA
232
GBP
EHERC10332000
Desisopropyl atrazine
Desisopropyl atrazine
Formula:
C₅H₈ClN₅ MW: 173.6 g/mol |
CAS Number:
1007-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...