Cyanuric acid, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3,5-Triazinetrion1,3,5-Triazine-2,4,6-triolIsocyanuric acid1,3,5-Triazinane-2,4,6-trione

M-Clarity™ quality level = MQ100

16614-250MGEA 30.4 GBP
16614-250MG
Cyanuric acid, Supelco®
Cyanuric acid
Formula: C₃H₃N₃O₃
MW: 129.08 g/mol
Density: 2.5 g/cm³ (20 °C)
MDL Number: MFCD00082990
CAS Number: 108-80-5
EINECS: 203-618-0
Merck Index: 13,02728

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR