Cyanuric acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1,3,5-Triazinetrion
, 1,3,5-Triazine-2,4,6-triol
, Isocyanuric acid
, 1,3,5-Triazinane-2,4,6-trione
EHERC11815000EA
94.4
GBP
EHERC11815000
Cyanuric acid
Cyanuric acid
Formula:
C₃H₃N₃O₃ MW: 129.08 g/mol |
MDL Number:
MFCD00082990 CAS Number: 108-80-5 EINECS: 203-618-0 Merck Index: 13,02728 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...