Ruthenium(IV) oxide, anhydrous ≥99.9% (Ru basis)

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ruthenium dioxideRuthenium tetroxide

STRE44-1977-5GEA 0 GBP
STRE44-1977-5G
Ruthenium(IV) oxide, anhydrous ≥99.9% (Ru basis)
Ruthenium (IV) oxide
Formula: O₂Ru
MW: 133.07 g/mol
Melting Pt: Dec.
Density: 6.97
MDL Number: MFCD00074857
CAS Number: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR