Iron(II) fluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ferrous fluoride, Iron difluoride
006158-10G
006158-25G
006158-100G
FLUO006158-10GEA
0
GBP
FLUO006158-10G
FLUO006158-25G
FLUO006158-100G
Iron(II) fluoride
Iron (II) fluoride
Formula:
F₂Fe MW: 93.84 g/mol Boiling Pt: 1100 °C (1013 hPa) Melting Pt: 965…975 °C Density: 4.09 g/cm³ (20 °C) |
MDL Number:
MFCD00016093 CAS Number: 7789-28-8 EINECS: 232-155-7 UN: 3288 ADR: 6.1,III Merck Index: 12,04093 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...