2-Acetylnaphthalene
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
Methyl β-naphthyl ketone
, 2'-Acetonaphthone
, 2´-Acetonaphthone
, 2-Acetonaphthone
, 1-(Naphthalen-2-yl)ethanone
MOLE82226206-100GEA
0
GBP
MOLE82226206-100G
2-Acetylnaphthalene
2-Acetylnaphthalene
Formula:
C₁₂H₁₀O MW: 170.21 g/mol Boiling Pt: 300 °C (1013 hPa) Melting Pt: 52…54 °C Density: 1.1171 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004108 CAS Number: 93-08-3 EINECS: 202-216-2 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...