Diisopropyl sulphide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Isopropyl sulphide

I26809-25G I26809-100G
SIALI26809-25GEA 135 GBP
SIALI26809-25G SIALI26809-100G
Diisopropyl sulphide, Sigma-Aldrich®
Diisopropyl sulphide
Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –69.1 °C
Density: 0,8306 g/cm³ (0 °C)
MDL Number: MFCD00008895
CAS Number: 625-80-9
EINECS: 210-911-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR