Tetrahexylammonium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

263834-25G 263834-5G
SIAL263834-25GEA 389 GBP
SIAL263834-25G SIAL263834-5G
Tetrahexylammonium chloride, Sigma-Aldrich®
Tetrahexylammonium chloride
Formula: C₂₄H₅₂ClN
MW: 390.14 g/mol
Melting Pt: 111…113 °C
CAS Number: 5922-92-9
EINECS: 227-653-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR