1,2-Bis(dimethylphosphino)ethane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethane-1,3-diylbis(dimethylphosphine), 1,2-Bis(dimethylphosphino)ethane, Bis(1,2-dimethylphosphino)ethane, DMPE, Ethylenebis(dimethylphosphine)

M-Clarity™ quality level = MQ100

261939-1G 261939-5G
SIAL261939-1GEA 209 GBP
SIAL261939-1G SIAL261939-5G
1,2-Bis(dimethylphosphino)ethane, Sigma-Aldrich®
1,2-Bis(dimethylphosphino)ethane
Formula: C₆H₁₆P₂
MW: 150.14 g/mol
Boiling Pt: 26 °C (1 mmHg)
Melting Pt: 147…149 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00008511
CAS Number: 23936-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR