Gallium(III) acetylacetonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ga(acac)3Gallium (III) tris(2,4-pentanedionato-O,O')Gallium(III) AcetylacetonateTris(2,4-pentanedionato)gallium(III)Gallium (III) 2,4-pentanedionateAcetylacetone Gallium(III) saltTris(2,4-pentanedionato-O,O')gallium (III)

M-Clarity™ quality level = MQ100

SIAL393541-5GEA 230 GBP
SIAL393541-5G SIAL393541-25G
Gallium(III) acetylacetonate, Sigma-Aldrich®
Gallium (III) acetylacetonate
Formula: C₁₅H₂₁GaO₆
MW: 367.05 g/mol
Melting Pt: 196…198 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00013492
CAS Number: 14405-43-7
EINECS: 238-377-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR