PIPES disodium salt
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Piperazine-N,N'-bis-2-ethanesulphonic acid disodium salt
, Disodium piperazine-1,4-diethanesulphonate
, 1,4-Piperazinediethanesulphonic acid disodium salt
, PIPES-Na2
, Piperazine-1,4-bis(2-ethanesulphonic acid) disodium salt
, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid disodium salt
ROTH7109.2EA
120.17
GBP
ROTH7109.2
PIPES disodium salt
PIPES disodium salt
Formula:
C₈H₁₆N₂Na₂O₆S₂ MW: 346.34 g/mol |
MDL Number:
MFCD00064349 CAS Number: 76836-02-7 EINECS: 278-562-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...