1-(3-Hydroxypropyl)piperazine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Piperazinepropanol, 3-Piperazin-1-ylpropanol, 3-(Piperazin-1-yl)propan-1-ol
641642-1G
641642-5G
SIAL641642-1GEA
63.8
GBP
SIAL641642-1G
SIAL641642-5G
1-(3-Hydroxypropyl)piperazine, Sigma-Aldrich®
1-(3-Hydroxypropyl)piperazine
Formula:
C₇H₁₆N₂O MW: 144.22 g/mol Boiling Pt: 264 °C (1013 hPa) Melting Pt: 49 °C |
MDL Number:
MFCD00023132 CAS Number: 5317-32-8 EINECS: 226-176-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...