4-Methoxybiphenyl

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Biphenyl-4-yl methyl ether, 4-Phenylanisole

018785-10G 018785-25G
FLUO018785-10GEA 0 GBP
FLUO018785-10G FLUO018785-25G
4-Methoxybiphenyl
4-Methoxybiphenyl
Formula: C₁₃H₁₂O
MW: 184.24 g/mol
MDL Number: MFCD00014897
CAS Number: 613-37-6
EINECS: 210-339-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR