2-Chlorobenzyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-2-(isothiocyanatomethyl)benzene

C1838.5 C1838.25
XXXXC1838.5EA 0 GBP
XXXXC1838.5 XXXXC1838.25
2-Chlorobenzyl isothiocyanate
2-Chlorobenzyl isothiocyanate
Formula: C₈H₆ClNS
Boiling Pt: 94…98 °C (0.2 mmHg)
Density: 1,282 g/cm³ (20 °C)
MDL Number: MFCD00041110
CAS Number: 18967-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR