2-Chlorobenzyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-2-(isothiocyanatomethyl)benzene

002527-5G 002527-1G
FLUO002527-5GEA 0 GBP
FLUO002527-5G FLUO002527-1G
2-Chlorobenzyl isothiocyanate
2-Chlorobenzyl isothiocyanate
Formula: C₈H₆ClNS
Boiling Pt: 94…98 °C (0,2 mmHg)
Density: 1,282 g/cm³ (20 °C)
MDL Number: MFCD00041110
CAS Number: 18967-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR