2-Chloro-4'-fluoroacetophenone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone
M-Clarity™ quality level = MQ100
132888-100G
132888-25G
SIAL132888-100GEA
606
GBP
SIAL132888-100G
SIAL132888-25G
2-Chloro-4'-fluoroacetophenone, Sigma-Aldrich®
2-Chloro-4'-fluoroacetophenone
Formula:
C₈H₆ClFO MW: 172.59 g/mol Boiling Pt: 247 °C (1013 hPa) Melting Pt: 45 °C |
MDL Number:
MFCD00011652 CAS Number: 456-04-2 EINECS: 207-256-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...