2-Chloro-4'-fluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-1-(4-fluorophenyl)ethan-1-one, α-Chloro-4-fluoroacetophenone, 2-Chloro-1-(4-fluorophenyl)ethanone, Chloromethyl p-fluorophenyl ketone, 4-Fluorophenacyl chloride, 2-Chloro-4'-fluoroacetophenone

001629-25G 001629-100G
FLUO001629-25GEA 0 GBP
FLUO001629-25G FLUO001629-100G
2-Chloro-4'-fluoroacetophenone
2-Chloro-4'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 45 °C
MDL Number: MFCD00011652
CAS Number: 456-04-2
EINECS: 207-256-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR