2',6'-Dichloro-3'-fluoroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009128-5G
009128-25G
009128-100G
FLUO009128-5GEA
0
GBP
FLUO009128-5G
FLUO009128-25G
FLUO009128-100G
2',6'-Dichloro-3'-fluoroacetophenone
2',6'-Dichloro-3'-fluoroacetophenone
Formula:
C₈H₅Cl₂FO MW: 207.03 g/mol |
MDL Number:
MFCD02093760 CAS Number: 290835-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...