ω,ω-Dichloroacetophenone ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2-Dichloroacetophenone, α,α-Dichloroacetophenone
B23475.06
B23475.14
B23475.06EA
0
GBP
B23475.06
B23475.14
ω,ω-Dichloroacetophenone ≥97%
ω,ω-Dichloroacetophenone
Purity: 97%
CAS No.: 2648-61-5
Molecular Formula: C8H6Cl2O
Formula Weight: 189.04
Flash Point:
Formula:
(C₆H₅)C(O)CHCl₂ MW: 189.04 g/mol Boiling Pt: 252 °C (1013 hPa) Melting Pt: 23…24 °C Density: 1.34 g/cm³ (16 °C) Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00000844 CAS Number: 2648-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...