2-Chloro-4-iodoaniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

652679-25G 652679-5G
SIAL652679-25GEA 199 GBP
SIAL652679-25G SIAL652679-5G
2-Chloro-4-iodoaniline, Sigma-Aldrich®
2-Chloro-4-iodoaniline
Formula: C₆H₅ClIN
MW: 253.47 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 73 °C
MDL Number: MFCD00025298
CAS Number: 42016-93-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR