3-Chloro-2-fluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005665-1G 005665-5G
FLUO005665-1GEA 0 GBP
FLUO005665-1G FLUO005665-5G
3-Chloro-2-fluorophenol
3-Chloro-2-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
MDL Number: MFCD01631572
CAS Number: 2613-22-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR