4-Bromo-2-(trifluoromethoxy)aniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

631256-25G 631256-5G
SIAL631256-25GEA 366 GBP
SIAL631256-25G SIAL631256-5G
4-Bromo-2-(trifluoromethoxy)aniline, Sigma-Aldrich®
4-Bromo-2-(trifluoromethoxy)aniline
Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
MDL Number: MFCD00179338
CAS Number: 175278-09-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR